2-(Methylthio)phenol ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-Hydroxythioanisole
M1854-5G
TCM1854-5GEA
110.64
USD
TCM1854-5G
2-(Methylthio)phenol ≥97.0% (by GC, titration analysis)
2-(Methylthio)phenol
CAS Number: 1073-29-6
MDL Number: MFCD00002211
Molecular Formula: C7H8OS
Molecular Weight: 140.20
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 105
Flash Point (°C): 110
Specific Gravity (20/20): 1.16
Formula:
C₇H₈OS MW: 140.21 g/mol Boiling Pt: 105 °C Density: 1.16 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00002211 CAS Number: 1073-29-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...