4-(Methylthio)thiophenol ≥97.0% (by titrimetric analysis)
Supplier: TCI America
M1823-5G
TCM1823-5GEA
146.3
USD
TCM1823-5G
4-(Methylthio)thiophenol ≥97.0% (by titrimetric analysis)
4-(Methylthio)thiophenol
CAS Number: 1122-97-0
MDL Number: MFCD00082770
Molecular Formula: C7H8S2
Molecular Weight: 156.26
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 117
Flash Point (°C): 110
Specific Gravity (20/20): 1.19
Other Product Names / Synonyms: 4-(Methylthio)thiophenol, 4-Mercaptothioanisole
Formula:
C₇H₈S₂ Boiling Pt: 117 °C Density: 1.19 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00082770 CAS Number: 1122-97-0 |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...