3-(Methylthio)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
3-(Methylthio)benzeneboronic acid
, 3-Thioanisoleboronic acid
M1793-5G
TCM1793-5GEA
109
USD
TCM1793-5G
3-(Methylthio)phenylboronic acid (contains varying amounts of Anhydride)
3-(Methylthio)phenylboronic acid
CAS Number: 128312-11-8
MDL Number: MFCD01319105
Molecular Formula: C7H9BO2S
Molecular Weight: 168.02
Form: Crystal
Color: White
Melting point (°C): 162
Other Product Names / Synonyms: 3-(Methylthio)benzeneboronic Acid
Formula:
C₇H₉BO₂S MW: 168.02 g/mol Melting Pt: 162 °C |
MDL Number:
MFCD01319105 CAS Number: 128312-11-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...