4'-Methylformanilide ≥98.0%

Supplier: TCI America

M1721-5G
TCM1721-5GEA 222.29 USD
TCM1721-5G
4'-Methylformanilide ≥98.0%
4'-Methylformanilide

CAS Number: 3085-54-9
Molecular Formula: C8H9NO
Molecular Weight: 135.17
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 53
Flash Point (°C): 110


Other Product Names / Synonyms: N-Formyl-p-toluidine, N-(p-Tolyl)formamide

Formula: C₈H₉NO
MW: 135.17 g/mol
Melting Pt: 53 °C
Flash Pt: 110 °C
CAS Number: 3085-54-9
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/60/45/28976045.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR