5-Methylhydantoin ≥98.0% (by titrimetric analysis)

M1699-1G
TCM1699-1GEA 29.97 USD
TCM1699-1G
5-Methylhydantoin ≥98.0% (by titrimetric analysis)
5-Methylhydantoin

CAS Number: 616-03-5
Molecular Formula: C4H6N2O2
Molecular Weight: 114.10
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 150

Formula: C₄H₆N₂O₂
Melting Pt: 150 °C
MDL Number: MFCD00022398
CAS Number: 616-03-5
https://us.vwr.com/stibo/web/std.lang.all/60/12/28976012.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR