2-(N-Methyl-2-pyridylamino)ethanol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-(Methyl(pyridin-2-yl)amino)ethanol
, 2-[N-(2-Hydroxyethyl)-N-methylamino]pyridine
, N-Methyl-N-(2-pyridyl)ethanolamine
M1468-5G
TCM1468-5GEA
73.08
USD
TCM1468-5G
2-(N-Methyl-2-pyridylamino)ethanol ≥98.0% (by GC, titration analysis)
2-(N-Methyl-2-pyridylamino)ethanol
CAS Number: 122321-04-4
MDL Number: MFCD02667633
Molecular Formula: C8H12N2O
Molecular Weight: 152.20
Purity/Analysis Method:
Form: Clear Liquid
Specific Gravity (20/20): 1.12
Other Product Names / Synonyms: 2-[N-(2-Hydroxyethyl)-N-methylamino]pyridine, N-Methyl-N-(2-pyridyl)ethanolamine
Formula:
C₈H₁₂N₂O MW: 152.20 g/mol Density: 1.12 (20 °C) |
MDL Number:
MFCD02667633 CAS Number: 122321-04-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...