1-(4-Methoxyphenyl)piperazine dihydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

M1449-5G M1449-25G
TCM1449-25GEA 112 USD
TCM1449-25G TCM1449-5G
1-(4-Methoxyphenyl)piperazine dihydrochloride ≥98.0% (by HPLC, titration analysis)
1-(4-Methoxyphenyl)piperazine dihydrochloride

CAS Number: 38869-47-5
MDL Number: MFCD00012767
Molecular Formula: C11H16N2O
Molecular Weight: 265.18
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 4-(1-Piperazinyl)anisole Dihydrochloride

Formula: C₁₁H₁₈Cl₂N₂O
MDL Number: MFCD00012767
CAS Number: 38869-47-5

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR