4'-Methoxybutyrophenone ≥98.0%
Supplier: TCI America
Synonyms:
1-(4-Methoxyphenyl)butan-1-one
M1436-5G
M1436-1G
TCM1436-5GEA
78.12
USD
TCM1436-5G
TCM1436-1G
4'-Methoxybutyrophenone ≥98.0%
4'-Methoxybutyrophenone
CAS Number: 4160-51-4
MDL Number: MFCD00027138
Molecular Formula: C11H14O2
Molecular Weight: 178.23
Purity/Analysis Method:
Form: Crystal
Freezing point (°C): 22
Other Product Names / Synonyms: p-Butyrylanisole
Formula:
C₁₁H₁₄O₂ MW: 178.23 g/mol |
MDL Number:
MFCD00027138 CAS Number: 4160-51-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...