6-Methoxyquinaldine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M1414-5G M1414-25G
TCM1414-25GEA 528.82 USD
TCM1414-25G TCM1414-5G
6-Methoxyquinaldine ≥98.0% (by GC, titration analysis)
6-Methoxyquinaldine

CAS Number: 1078-28-0
MDL Number: MFCD00006761
Molecular Formula: C11H11NO
Molecular Weight: 173.22
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 67


Other Product Names / Synonyms: 6-Methoxyquinaldine

Formula: C₁₁H₁₁NO
MW: 173.21 g/mol
Melting Pt: 67 °C
MDL Number: MFCD00006761
CAS Number: 1078-28-0
https://us.vwr.com/stibo/web/std.lang.all/55/56/28975556.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR