7-Methoxy-4-methylcoumarin ≥98.0%

Supplier: TCI America

M1393-5G
TCM1393-5GEA 110.6 USD
TCM1393-5G
7-Methoxy-4-methylcoumarin ≥98.0%
7-Methoxy-4-methylcoumarin

CAS Number: 2555-28-4
MDL Number: MFCD00009773
Molecular Formula: C11H10O3
Molecular Weight: 190.20
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 160

Formula: C₁₁H₁₀O₃
MW: 190.2 g/mol
Melting Pt: 160 °C
MDL Number: MFCD00009773
CAS Number: 2555-28-4
https://us.vwr.com/stibo/web/std.lang.all/55/17/28975517.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR