2-Mercapto-5-methylthio-1,3,4-thiadiazole ≥98.0% (by HPLC, titration analysis)
Synonyms:
5-Methylthio-1,3,4-thiadiazole-2-thiol
, 5-Methylthio-1,3,4-thiadiazole-2(3H)-thione
, 5-(Methylthio)-1,3,4-thiadiazole-2-thiol
M1384-25G
TCM1384-25GEA
128.76
USD
TCM1384-25G
2-Mercapto-5-methylthio-1,3,4-thiadiazole ≥98.0% (by HPLC, titration analysis)
2-Mercapto-5-methylthio-1,3,4-thiadiazole
CAS Number: 6264-40-0
MDL Number: MFCD00051648
Molecular Formula: C3H4N2S3
Molecular Weight: 164.26
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 137
Other Product Names / Synonyms: 5-Methylthio-1,3,4-thiadiazole-2-thiol, 5-Methylthio-1,3,4-thiadiazole-2(3H)-thione
Formula:
C₃H₄N₂S₃ MW: 164.26 g/mol Melting Pt: 137 °C |
MDL Number:
MFCD00051648 CAS Number: 6264-40-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...