3-Methyl-4-nitroanisole ≥98.0%

Supplier: TCI America

Synonyms: Methyl 4-nitro-m-tolyl ether4-Methoxy-2-methyl-1-nitrobenzene5-Methoxy-2-nitrotoluene1-Methoxy-3-methyl-4-nitrobenzene

M1369-25G
TCM1369-25GEA 55.83 USD
TCM1369-25G
3-Methyl-4-nitroanisole ≥98.0%
3-Methyl-4-nitroanisole

CAS Number: 5367-32-8
MDL Number: MFCD00007167
Molecular Formula: C8H9NO3
Molecular Weight: 167.16
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 135
Melting point (°C): 51
Flash Point (°C): 230


Other Product Names / Synonyms: 1-Methoxy-3-methyl-4-nitrobenzene, 3-Methyl-4-nitroanisole

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Boiling Pt: 135 °C
Melting Pt: 51 °C
Flash Pt: 230 °C
MDL Number: MFCD00007167
CAS Number: 5367-32-8
UN: 2446
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/54/33/28975433.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR