3-(4-Methoxyphenoxy)benzaldehyde ≥97.0%
Supplier: TCI America
M1361-1G
TCM1361-001GEA
262
USD
TCM1361-001G
3-(4-Methoxyphenoxy)benzaldehyde ≥97.0%
3-(4-Methoxyphenoxy)benzaldehyde
CAS Number: 62373-80-2
MDL Number: MFCD00003358
Molecular Formula: C14H12O3
Molecular Weight: 228.25
Purity/Analysis Method:
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.19
Other Product Names / Synonyms: 3-Formyl-4'-methoxydiphenyl Ether
Formula:
C₁₄H₁₂O₃ Density: 1.19 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00003358 CAS Number: 62373-80-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...