3-Mercaptopropyl(dimethoxy)methylsilane ≥95.0%
Supplier: TCI America
Synonyms:
3-(Dimethoxymethylsilyl)-1-propanethiol
CAS Number: 31001-77-1
MDL Number: MFCD00053923
Molecular Formula: C6H16O2SSi
Molecular Weight: 180.34
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 74
Flash Point (°C): 92
Specific Gravity (20/20): 1.00
Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents, Silane Coupling Agents
Other Product Names / Synonyms: 3-(Dimethoxymethylsilyl)-1-propanethiol, 3-Mercaptopropylmethyldimethoxysilane
Formula:
C₆H₁₆O₂SSi MW: 180.34 g/mol Boiling Pt: 74 °C Density: 1.00 (20 °C) Flash Pt: 92 °C |
MDL Number:
MFCD00053923 CAS Number: 31001-77-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...