Tetraisopropyl methylenebisphosphonate ≥97.0%
Supplier: TCI America
Synonyms:
Tetraisopropyl methylenebis(phosphonate)
, Tetraisopropyl methylenediphosphonate
CAS Number: 1660-95-3
MDL Number: MFCD00015021
Molecular Formula: C13H30O6P2
Molecular Weight: 344.32
Purity/Analysis Method:
Form: Clear Liquid
Flash Point (°C): 160
Specific Gravity (20/20): 1.06
Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: Methylenediphosphonic Acid Tetraisopropyl Ester
Formula:
C₁₃H₃₀O₆P₂ MW: 344.33 Density: 1.06 (20 °C) Flash Pt: 160 °C |
MDL Number:
MFCD00015021 CAS Number: 1660-95-3 UN: 3278 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...