Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(Methylamino)-1-(methylthio)-2-nitroethene ≥99.0% (by HPLC)

Supplier: TCI America

Synonyms: 1-Methylthio-1-methylamino-2-nitroethylene

M1307-25G M1307-500G
TCM1307-025GEA 54.11 USD InStock
TCM1307-025G TCM1307-500G
1-(Methylamino)-1-(methylthio)-2-nitroethene ≥99.0% (by HPLC)
1-(Methylamino)-1-(methylthio)-2-nitroethene
CAS Number: 61832-41-5
MDL Number: MFCD00040994
Molecular Formula: C4H8N2O2S
Molecular Weight: 148.18
Purity/Analysis Method: 99.0% (HPLC)
Form: Crystal
Melting point (°C): 112

Other Product Names / Synonyms: N-Methyl-1-methylthio-2-nitroethenamine


Formula: C₄H₈N₂O₂S
MW: 148,18
Melting Pt: 112 °C
MDL Number: MFCD00040994
CAS Number: 61832-41-5
EINECS: 263-266-9
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (HPLC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty