Methyltrioxorhenium(VII) ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: MTOMethylrhenium Trioxide(VII)

M1296-100MG
TCM1296-100MGEA 37 USD
TCM1296-100MG
Methyltrioxorhenium(VII) ≥97.0% (by titrimetric analysis)
Methyltrioxorhenium(VII)

CAS Number: 70197-13-6
MDL Number: MFCD00192332
Molecular Formula: CH3O3Re
Molecular Weight: 249.24
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Melting point (°C): 114


Other Product Names / Synonyms: Methylrhenium Trioxide(VII), MTO

Formula: CH₃O₃Re
MW: 249.24 g/mol
Melting Pt: 114 °C
MDL Number: MFCD00192332
CAS Number: 70197-13-6
https://us.vwr.com/stibo/web/std.lang.all/51/70/28975170.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR