2-Mercapto-5-methoxybenzothiazole ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
5-Methoxybenzo[d]thiazole-2-thiol
M1277-5G
TCM1277-005GEA
231.36
USD
TCM1277-005G
2-Mercapto-5-methoxybenzothiazole ≥98.0% (by titrimetric analysis)
2-Mercapto-5-methoxybenzothiazole
CAS Number: 55690-60-3
MDL Number: MFCD00185941
Molecular Formula: C8H7NOS2
Molecular Weight: 197.27
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 198
Other Product Names / Synonyms: 5-Methoxybenzothiazole-2-thiol
Formula:
C₈H₇NOS₂ MW: 197.28 g/mol Melting Pt: 198 °C |
MDL Number:
MFCD00185941 CAS Number: 55690-60-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...