2-Methoxy-6-(methylamino)pyridine ≥98.0% (by GC, titration analysis)
M1185-25G
TCM1185-025GEA
130.81
USD
TCM1185-025G
2-Methoxy-6-(methylamino)pyridine ≥98.0% (by GC, titration analysis)
2-Methoxy-6-(methylamino)pyridine
CAS Number: 88569-83-9
MDL Number: MFCD00191557
Molecular Formula: C7H10N2O
Molecular Weight: 138.17
Purity/Analysis Method:
Form: Clear Liquid
Specific Gravity (20/20): 1.09
Storage Temperature:
Formula:
C₇H₁₀N₂O MW: 138.17 g/mol Density: 1.09 (20 °C) |
MDL Number:
MFCD00191557 CAS Number: 88569-83-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...