TCIAM1131

M1131-25G
TCM1131-025GEA 199.85 USD
TCM1131-025G
TCIAM1131
CAS Number: 2786-31-4
MDL Number: MFCD00050297
Molecular Formula: C8H8INS
Molecular Weight: 277.12
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR