4-Methyl-3-nitrophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
4-Methyl-3-nitrobenzeneboronic acid
, (4-Methyl-3-nitrophenyl)boronic acid
M1127-5G
M1127-1G
TCM1127-1GEA
115.71
USD
TCM1127-1G
TCM1127-005G
4-Methyl-3-nitrophenylboronic acid (contains varying amounts of Anhydride)
4-Methyl-3-nitrophenylboronic acid
CAS Number: 80500-27-2
MDL Number: MFCD00191550
Molecular Formula: C7H8BNO4
Molecular Weight: 180.95
Form: Crystal
Color: Very Pale Yellow
Other Product Names / Synonyms: 3-Nitro-p-tolylboronic Acid
Formula:
C₇H₈BNO₄ MW: 180.96 g/mol |
MDL Number:
MFCD00191550 CAS Number: 80500-27-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...