Ethyl-2-mercaptopropionate ≥97.0%

Supplier: TCI America

Synonyms: Ethyl 2-mercaptopropanoate2-Mercaptopropionic Acid Ethyl Ester

M1037-25G
TCM1037-025GEA 115 USD
TCM1037-025G
Ethyl-2-mercaptopropionate ≥97.0%
Ethyl-2-mercaptopropionate

CAS Number: 19788-49-9
MDL Number: MFCD00040233
Molecular Formula: C5H10O2S
Molecular Weight: 134.19
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 58
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: 2-Mercaptopropionic Acid Ethyl Ester

Formula: C₅H₁₀O₂S
MW: 134.19 g/mol
Boiling Pt: 90 °C
Density: 1.04 (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00040233
CAS Number: 19788-49-9
https://us.vwr.com/stibo/web/std.lang.all/46/62/28974662.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR