Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Mercapto-5-methyl-1,3,4-thiadiazole ≥98.0% (by titrimetric analysis)

Supplier: TCI America


M0976-250G M0976-25G
TCM0976-250GEA 210.14 USD InStock
TCM0976-250G TCM0976-025G
2-Mercapto-5-methyl-1,3,4-thiadiazole ≥98.0% (by titrimetric analysis)
2-Mercapto-5-methyl-1,3,4-thiadiazole
CAS Number: 29490-19-5
MDL Number: MFCD00038349
Molecular Formula: C3H4N2S2
Molecular Weight: 132.20
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 186

Other Product Names / Synonyms: 2-Mercapto-5-methyl-1,3,4-thiadiazole


Formula: C₃H₄N₂S₂
MW: 132,21
Melting Pt: 186 °C
MDL Number: MFCD00038349
CAS Number: 29490-19-5
EINECS: 249-667-1
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty