Methyl (methylsulfinyl)methyl sulfide ≥96.0%

Synonyms: Formaldehyde Dimethyl Dithioacetal S-OxideMethyl((methylsulfinyl)methyl)sulfaneMethyl (Methylthio)methyl sulphoxideFAMSOMethyl((methylsulphinyl)methyl)sulphaneMethyl (Methylthio)methyl sulfoxide

M0805-5G M0805-25G
TCM0805-5GEA 36.05 USD
TCM0805-5G TCM0805-25G
Methyl (methylsulfinyl)methyl sulfide ≥96.0%
Methyl (methylsulfinyl)methyl sulfide

CAS Number: 33577-16-1
MDL Number: MFCD00002091
Molecular Formula: C3H8OS2
Molecular Weight: 124.22
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 93
Flash Point (°C): 136
Specific Gravity (20/20): 1.22


Product Categories: Synthetic Organic Chemistry, Sulfoxides
Other Product Names / Synonyms: FAMSO, Formaldehyde Dimethyl Dithioacetal S-Oxide, Methyl (Methylthio)methyl Sulfoxide

Formula: C₃H₈OS₂
MW: 124.22 g/mol
Boiling Pt: 93 °C
Density: 1.22 (20 °C)
Flash Pt: 136 °C
MDL Number: MFCD00002091
CAS Number: 33577-16-1
https://us.vwr.com/stibo/web/std.lang.all/42/75/28974275.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR