2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid) ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2,2-Dimethyl-1,3-dioxane-4,6-dione

M0799-25G M0799-500G M0799-100G
TCM0799-500GEA 355.3 USD
TCM0799-500G TCM0799-100G TCM0799-025G
2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid) ≥98.0% (by titrimetric analysis)
2,2-Dimethyl-1,3-dioxane-4,6-dione (Isopropylidene malonate, Meldrum's acid)

CAS Number: 2033-24-1
MDL Number: MFCD00006638
Molecular Formula: C6H8O4
Molecular Weight: 144.13
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 95
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction)
Other Product Names / Synonyms: 2,2-Dimethyl-1,3-dioxane-4,6-dione, cycl-Isopropylidene Malonate, Malonic Acid Cyclic Isopropylidene Ester

Formula: C₆H₈O₄
MW: 144.13 g/mol
Melting Pt: 95 °C
MDL Number: MFCD00006638
CAS Number: 2033-24-1
https://us.vwr.com/stibo/web/std.lang.all/42/70/28974270.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR