Methylmagnesium bromide 35% ∼3 mol/L in ethyl ether
M0785-250G
TCM0785-250GEA
159.72
USD
TCM0785-250G
Methylmagnesium bromide 35% ∼3 mol/L in ethyl ether
Methylmagnesium bromide
CAS Number: 75-16-1
MDL Number: MFCD00000041
Molecular Formula: CH3BrMg
Molecular Weight: 119.24
Flash Point (°C): -40
Formula:
CH₃BrMg Flash Pt: –40 °C |
MDL Number:
MFCD00000041 CAS Number: 75-16-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...