4-Methylesculetin ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
6,7-Dihydroxy-4-methylcoumarin
M0766-25G
TCM0766-025GEA
85
USD
TCM0766-025G
4-Methylesculetin ≥98.0% (by HPLC, titration analysis)
4-Methylesculetin
CAS Number: 529-84-0
MDL Number: MFCD00006859
Molecular Formula: C10H8O4
Molecular Weight: 192.17
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 278
Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: 6,7-Dihydroxy-4-methylcoumarin
Formula:
C₁₀H₈O₄ MW: 192.17 g/mol Melting Pt: 278 °C |
MDL Number:
MFCD00006859 CAS Number: 529-84-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...