2-Methyl-3-(methylthio)pyrazine ≥99.0%
M0685-1ML
TCM0685-001MLEA
56.65
USD
TCM0685-001ML
2-Methyl-3-(methylthio)pyrazine ≥99.0%
2-Methyl-3-(methylthio)pyrazine
CAS Number: 2882-20-4
MDL Number: MFCD00014610
Molecular Formula: C6H8N2S
Molecular Weight: 140.20
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 104
Flash Point (°C): 76
Specific Gravity (20/20): 1.15
Other Product Names / Synonyms: 2-Methylmercapto-3-methylpyrazine
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...