4-Methyl-2-phenylimidazole ≥93.0%

Supplier: TCI America

Synonyms: 4-Methyl-2-phenyl-1H-imidazole

M0669-25G
TCM0669-025GEA 48 USD
TCM0669-025G
4-Methyl-2-phenylimidazole ≥93.0%
4-Methyl-2-phenylimidazole

CAS Number: 827-43-0
MDL Number: MFCD00047015
Molecular Formula: C10H10N2
Molecular Weight: 158.20
Purity/Analysis Method: 93.0% (GC)
Form: Crystal
Melting point (°C): 181
Flash Point (°C): 220

Formula: C₁₀H₁₀N₂
MW: 158.20 g/mol
Melting Pt: 181 °C
Flash Pt: 220 °C
MDL Number: MFCD00047015
CAS Number: 827-43-0
https://us.vwr.com/stibo/web/std.lang.all/41/39/28974139.jpg

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR