Methylsulfonylacetonitrile ≥98.0%
Supplier: TCI America
Synonyms:
Methanesulphonylacetonitrile
, Methanesulfonylacetonitrile
, Cyanomethylsulphonylmethane
, Cyanomethylsulfonylmethane
, 2-(Methylsulphonyl)acetonitrile
, 2-(Methylsulfonyl)acetonitrile
CAS Number: 2274-42-2
MDL Number: MFCD00014743
Molecular Formula: C3H5NO2S
Molecular Weight: 119.14
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 84
Other Product Names / Synonyms: Cyanomethylsulfonylmethane, Methanesulfonylacetonitrile
Formula:
C₃H₅NO₂S MW: 119.14 g/mol Melting Pt: 84 °C |
MDL Number:
MFCD00014743 CAS Number: 2274-42-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...