3,4-Methylenedioxycinnamic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3-(1,3-Benzodioxol-5-yl)acrylic Acid3-Piperonylacrylic Acid

M0634-25G
TCM0634-025GEA 93.38 USD
TCM0634-025G
3,4-Methylenedioxycinnamic acid ≥98.0% (by HPLC, titration analysis)
3,4-Methylenedioxycinnamic acid

CAS Number: 2373-80-0
MDL Number: MFCD00005837
Molecular Formula: C10H8O4
Molecular Weight: 192.17
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 243


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: 3-(1,3-Benzodioxol-5-yl)acrylic Acid, 3-Piperonylacrylic Acid

Formula: C₁₀H₈O₄
MW: 192.17 g/mol
Melting Pt: 243 °C
MDL Number: MFCD00005837
CAS Number: 2373-80-0
https://us.vwr.com/stibo/web/std.lang.all/40/96/28974096.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR