TCIAM0625

M0625-1G
TCM0625-001GEA 92.41 USD
TCM0625-001G
TCIAM0625
CAS Number: 50257-39-1
MDL Number: MFCD00005284
Molecular Formula: C12H14N2O2S
Molecular Weight: 250.32
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Melting point (°C): 98

Product Categories: Synthetic Organic Chemistry, Nucleic Acid Synthesis
Other Product Names / Synonyms: 1-(2,4,6-Trimethylbenzenesulfonyl)imidazole, MSI
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR