Mucochloric acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,3-Dichloromalealdehydic acid

M0472-25G M0472-500G
TCM0472-25GEA 23 USD
TCM0472-25G TCM0472-500G
Mucochloric acid ≥98.0% (by GC, titration analysis)
Mucochloric acid

CAS Number: 87-56-9
MDL Number: MFCD00006966
Molecular Formula: C4H2Cl2O3
Molecular Weight: 168.96
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 127
Flash Point (°C): 100
Storage Temperature: 0-10°C

Formula: C₄H₂Cl₂O₃
MW: 168.96
Melting Pt: 127 °C
Flash Pt: 100 °C
MDL Number: MFCD00135465
UN: 2923
ADR: 8,III
https://us.vwr.com/stibo/web/std.lang.all/39/14/28973914.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR