D-(-)-Lyxose ≥98.0% (by HPLC)
Supplier: TCI America
L0073-1G
L0073-25G
L0073-5G
TCL0073-001GEA
46.69
USD
TCL0073-001G
TCL0073-25G
TCL0073-005G
D-(-)-Lyxose ≥98.0% (by HPLC)
D(-)-Lyxose
CAS Number: 1114-34-7
MDL Number: MFCD00064362
Molecular Formula: C5H10O5
Molecular Weight: 150.13
Purity/Analysis Method:
Form: Crystal
Specific rotation [a]20/D: -14 deg (C=1, H2O)
Formula:
C₅H₁₀O₅ MW: 150.13 |
MDL Number:
MFCD00064362 CAS Number: 1114-34-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...