Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iodoacetamide ≥98.0% (by total nitrogen basis), for Biochemical research

Supplier: TCI America

Synonyms: 2-Iodoacetamide

I0741-5G
TCI0741-5GEA 20.23 USD InStock
TCI0741-5G
Iodoacetamide ≥98.0% (by total nitrogen basis), for Biochemical research
Iodoacetamide
[for Biochemical Research]
CAS Number: 144-48-9
MDL Number: MFCD00008028
Molecular Formula: C2H4INO
Molecular Weight: 184.96
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Melting point (°C): 95


Formula: ICH₂CONH₂
MW: 184,96 g/mol
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 95 °C
Storage Temperature: 2 - 8°C
MDL Number: MFCD00008028
CAS Number: 144-48-9
EINECS: 205-630-1
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (N)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty