TCIAI0688

I0688-1G
TCI0688-1GEA 28.1 USD
TCI0688-1G
TCIAI0688
CAS Number: 20513-98-8
MDL Number: MFCD00061641
Molecular Formula: C9H12O6
Molecular Weight: 216.19
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 121
Specific rotation [a]20/D: 67 deg (C=1, H2O)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR