Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Hydroxy-4-methoxybenzyl alcohol ≥98.0%

Supplier: TCI America


I0648-5G
TCI0648-5GEA 18.05 USD InStock
TCI0648-5G
3-Hydroxy-4-methoxybenzyl alcohol ≥98.0%
3-Hydroxy-4-methoxybenzyl alcohol
CAS Number: 4383-06-6
Molecular Formula: C8H10O3
Molecular Weight: 154.17
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 136

Other Product Names / Synonyms: 3-Hydroxy-4-methoxybenzyl Alcohol


Formula: C₈H₁₀O₃
MW: 154,17 g/mol
Melting Pt: 136 °C
MDL Number: MFCD00004644
CAS Number: 4383-06-6
EINECS: 224-489-7
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty