Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Iodoisatin ≥98.0% (by titrimetric analysis)

Supplier: TCI America


I0640-5G
TCI0640-5GEA 76.99 USD InStock
TCI0640-5G
5-Iodoisatin ≥98.0% (by titrimetric analysis)
5-Iodoisatin
CAS Number: 20780-76-1
Molecular Formula: C8H4INO2
Molecular Weight: 273.03
Purity/Analysis Method: 98.0% (T)
Form: Crystal

Product Categories: Synthetic Organic Chemistry, Isatins (Multi-component Reaction)
Other Product Names / Synonyms: 5-Iodoindole-2,3-dione, 5-Iodo-2,3-indolinedione


Formula: C₈H₄INO₂
MW: 273,03 g/mol
MDL Number: MFCD00016899
CAS Number: 20780-76-1
EINECS: 244-035-1
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty