Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isobornyl Acrylate ≥90.0% stabilized

Supplier: TCI America

Synonyms: Acrylic Acid Isobornyl Ester

I0638-500G I0638-25G
TCI0638-500GEA 95.68 USD InStock
TCI0638-500G TCI0638-25G
Isobornyl Acrylate ≥90.0% stabilized
Isobornyl acrylate
(stabilized with MEHQ)
CAS Number: 5888-33-5
MDL Number: MFCD00080424
Molecular Formula: C13H20O2
Molecular Weight: 208.30
Purity/Analysis Method: 90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 104
Flash Point (°C): 109
Specific Gravity (20/20): 0.99

Product Categories: Materials Chemistry, Monomers, Photopolymer Reagents, Bioscience, Bicyclic Monoterpenes
Other Product Names / Synonyms: Acrylic Acid Isobornyl Ester


Formula: C₁₃H₂₀O₂
MW: 208,30 g/mol
Boiling Pt: 104 °C
Density: 0,99 (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD00080424
CAS Number: 5888-33-5
Order Now
Specification Test Results
Purity/Analysis Method 90.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty