Methyl isobutyrylacetate ≥97.0%
Supplier: TCI America
Synonyms:
Methyl 4-methyl-3-oxopentanoate
I0592-25G
TCI0592-25GEA
65
USD
TCI0592-25G
Methyl isobutyrylacetate ≥97.0%
Methyl isobutyrylacetate
CAS Number: 42558-54-3
MDL Number: MFCD00040499
Molecular Formula: C7H12O3
Molecular Weight: 144.17
Purity/Analysis Method:
Form: Clear Liquid
Flash Point (°C): 78
Specific Gravity (20/20): 1.01
Other Product Names / Synonyms: Isobutyrylacetic Acid Methyl Ester, Methyl 4-Methyl-3-oxovalerate, 4-Methyl-3-oxovaleric Acid Methyl Ester
Formula:
C₇H₁₂O₃ MW: 144.17 g/mol Density: 1.01 (20 °C) Flash Pt: 78 °C |
MDL Number:
MFCD00040499 CAS Number: 42558-54-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...