Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl 2-isothiocyanatobenzoate ≥98.0% (by GC, titration analysis)

Supplier: TCI America


I0574-25G
TCI0574-25GEA 93.64 USD InStock
TCI0574-25G
Methyl 2-isothiocyanatobenzoate ≥98.0% (by GC, titration analysis)
Methyl 2-isothiocyanatobenzoate
CAS Number: 16024-82-1
MDL Number: MFCD00041062
Molecular Formula: C9H7NO2S
Molecular Weight: 193.22
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 126

Other Product Names / Synonyms: 2-Isothiocyanatobenzoic Acid Methyl Ester


Formula: C₉H₇NO₂S
MW: 193.23 g/mol
Boiling Pt: 104 °C (1013 hPa)
Density: 1.25 g/cm³ (20 °C)
Flash Pt: 126 °C
MDL Number: MFCD00041062
CAS Number: 16024-82-1
EINECS: 240-164-2
UN: 2923
ADR: 8,III
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty