Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isoquinolin-N-oxide

Supplier: TCI America


I0540-5G I0540-25G
TCI0540-005GEA 129.01 USD InStock
TCI0540-005G TCI0540-025G
Isoquinolin-N-oxide
Isoquinolin-N-oxide
CAS Number: 1532-72-5
MDL Number: MFCD00006903
Molecular Formula: C9H7NO
Molecular Weight: 145.16
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 104


Formula: C₉H₇NO
MW: 145,16
Melting Pt: 104 °C
MDL Number: MFCD00006903
CAS Number: 1532-72-5
EINECS: 216-242-7
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty