Isochroman ≥98.0%

Supplier: TCI America

Synonyms: 3,4-Dihydro-1H-2-benzopyran

I0514-25G
TCI0514-025GEA 61.6 USD
TCI0514-025G
Isochroman ≥98.0%
Isochroman

CAS Number: 493-05-0
MDL Number: MFCD00006913
Molecular Formula: C9H10O
Molecular Weight: 134.18
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 66
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: 3,4-Dihydro-1H-2-benzopyran

Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 110 °C
Density: 1.07 (20 °C)
Flash Pt: 66 °C
MDL Number: MFCD00006913
CAS Number: 493-05-0
https://us.vwr.com/stibo/web/std.lang.all/23/76/28972376.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR