(S)-(+)-1-(1-Naphthyl)ethyl isocyanate ≥95.0%

Supplier: TCI America

I0398-5G I0398-1G
TCI0398-5GEA 398.9 USD
TCI0398-5G TCI0398-001G
(S)-(+)-1-(1-Naphthyl)ethyl isocyanate ≥95.0%
(S)-(+)-1-(1-Naphthyl)ethyl isocyanate

CAS Number: 73671-79-1
MDL Number: MFCD00064181
Molecular Formula: C13H11NO
Molecular Weight: 197.24
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 93
Specific Gravity (20/20): 1.13
Specific rotation [a]20/D: 45.5 deg (C=5, Toluene)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Optical Resolving Agents, Chiral Building Blocks, Analytical Science, e.e. Determination (HPLC Labeling Reagents), e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: Isocyanic Acid (S)-(+)-1-(1-Naphthyl)ethyl Ester

Formula: C₁₃H₁₁NO
Density: 1.13 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00064181
CAS Number: 73671-79-1
https://us.vwr.com/stibo/web/std.lang.all/22/33/28972233.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR