Butyl Isocyanatoacetate ≥95.0%

Supplier: TCI America

Synonyms: Isocyanatoacetic Acid Butyl Ester

I0358-5G I0358-25G
TCI0358-005GEA 130.13 USD
TCI0358-005G TCI0358-25G
Butyl Isocyanatoacetate ≥95.0%
Butyl Isocyanatoacetate

CAS Number: 17046-22-9
MDL Number: MFCD00041743
Molecular Formula: C7H11NO3
Molecular Weight: 157.17
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 79
Flash Point (°C): 88
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: Isocyanatoacetic Acid Butyl Ester

Formula: C₇H₁₁NO₃
MW: 157.17 g/mol
Boiling Pt: 79 °C
Density: 1.06 (20 °C)
Flash Pt: 88 °C
MDL Number: MFCD00041743
CAS Number: 17046-22-9
https://us.vwr.com/stibo/web/std.lang.all/21/84/28972184.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR