2-(Isopropylthio)ethanol ≥97.0%
Supplier: TCI America
Synonyms:
2-Hydroxyethyl Isopropyl Sulphide
, 2-Hydroxyethyl Isopropyl Sulfide
I0265-25ML
TCI0265-025MLEA
33.47
USD
TCI0265-025ML
2-(Isopropylthio)ethanol ≥97.0%
2-(Isopropylthio)ethanol
CAS Number: 40811-49-2
MDL Number: MFCD00014038
Molecular Formula: C5H12OS
Molecular Weight: 120.21
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 96
Flash Point (°C): 70
Specific Gravity (20/20): 0.98
Other Product Names / Synonyms: 2-Hydroxyethyl Isopropyl Sulfide
Formula:
C₅H₁₂OS MW: 120.21 g/mol Boiling Pt: 96 °C Density: 0.98 (20 °C) Flash Pt: 70 °C |
MDL Number:
MFCD00014038 CAS Number: 40811-49-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...