4-Isopropylbiphenyl

Supplier: TCI America

Synonyms: 4-Phenylcumene

I0171-5G I0171-25G
TCI0171-5GEA 152 USD
TCI0171-5G TCI0171-025G
4-Isopropylbiphenyl
4-Isopropylbiphenyl

CAS Number: 7116-95-2
MDL Number: MFCD00221718
Molecular Formula: C15H16
Molecular Weight: 196.29
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 291
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: 4-Phenylcumene

Formula: C₁₅H₁₆
MW: 196.29 g/mol
Boiling Pt: 291 °C
Density: 0.99 (20 °C)
MDL Number: MFCD00221718
CAS Number: 7116-95-2
https://us.vwr.com/stibo/web/std.lang.all/19/62/28971962.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR