Isobutyramide ≥98.0%

Supplier: TCI America

I0102-25G I0102-500G
TCI0102-025GEA 144 USD
TCI0102-025G TCI0102-500G
Isobutyramide ≥98.0%
Isobutyramide

CAS Number: 563-83-7
MDL Number: MFCD00008019
Molecular Formula: C4H9NO
Molecular Weight: 87.12
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 128

Formula: C₄H₉NO
MW: 87.12 g/mol
Melting Pt: 128 °C
MDL Number: MFCD00008019
CAS Number: 563-83-7
https://us.vwr.com/stibo/web/std.lang.all/18/87/28971887.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR