1-(6-Methoxy-2-naphthyl)ethanol ≥98.0%

Supplier: TCI America

Synonyms: 1-(6-Methoxynaphthalen-2-yl)ethanolDL-6-Methoxy-α-methyl-2-naphthalenemethanol

H1270-5G
TCH1270-5GEA 95 USD
TCH1270-5G
1-(6-Methoxy-2-naphthyl)ethanol ≥98.0%
1-(6-Methoxy-2-naphthyl)ethanol

CAS Number: 77301-42-9
MDL Number: MFCD01632581
Molecular Formula: C13H14O2
Molecular Weight: 202.25
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 113


Other Product Names / Synonyms: 6-Methoxy-alpha-methyl-2-naphthalenemethanol, 1-(6-Methoxy-2-naphthyl)ethanol

Formula: C₁₃H₁₄O₂
MW: 202.25 g/mol
Melting Pt: 113 °C
MDL Number: MFCD01632581
CAS Number: 77301-42-9
https://us.vwr.com/stibo/web/std.lang.all/11/91/28971191.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR