4-Hydroxyphenylacetone ≥97.0% (by GC, titration analysis)
Supplier: TCI America
H1249-5G
H1249-25G
TCH1249-5GEA
59.17
USD
TCH1249-5G
TCH1249-25G
4-Hydroxyphenylacetone ≥97.0% (by GC, titration analysis)
4-Hydroxyphenylacetone
CAS Number: 770-39-8
MDL Number: MFCD00210439
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 177
Melting point (°C): 40
Other Product Names / Synonyms: 1-(4-Hydroxyphenyl)-2-propanone
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...